5-Fluoro-3-pyridineboronic acid pinacol ester ≥95%
Supplier: Matrix Scientific
Synonyms:
3-Fluoropyridine-5-boronic acid pinacol ester, 3-Fluoro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
070316-1G
070316-5G
101944-226EA
172.31
USD
101944-226
101944-228
5-Fluoro-3-pyridineboronic acid pinacol ester ≥95%
5-Fluoro-3-pyridineboronic acid pinacol ester
Hazard code(s): Irritant
Formula:
C₁₁H₁₅BFNO₂ MW: 223.05 g/mol |
MDL Number:
MFCD07780754 CAS Number: 719268-92-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...