1-Boc-6-bromoindole ≥98%

Supplier: Matrix Scientific

Synonyms: tert-Butyl 6-bromo-1H-indole-1-carboxylate

070166-1G 070166-5G
101943-618EA 113.85 USD
101943-618 101943-620
1-Boc-6-bromoindole ≥98%
1-Boc-6-bromoindole

Hazard code(s): Irritant

Formula: C₁₃H₁₄BrNO₂
MW: 296.16 g/mol
MDL Number: MFCD09954826
CAS Number: 147621-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR