2,5-Dichloro-4-pyridineboronic acid ≥96%
Supplier: Matrix Scientific
Synonyms:
2,5-Dichloropyridine-4-boronic acid, 2,5-Dichloro-4-pyridylboronic acid
070164-1G
070164-5G
101943-608EA
786.15
USD
101943-608
101943-610
2,5-Dichloro-4-pyridineboronic acid ≥96%
2,5-Dichloro-4-pyridineboronic acid
Hazard code(s): Irritant
Formula:
C₅H₄BCl₂NO₂ MW: 191.81 g/mol |
MDL Number:
MFCD06798254 CAS Number: 847664-64-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...