4-Fluoroindole ≥99%

Supplier: Matrix Scientific
Warning

Synonyms: 4-Fluoro-1H-indole

069738-1G 069738-5G
101941-702EA 35.38 USD
101941-702 101941-704
4-Fluoroindole ≥99%
4-Fluoroindole

Hazard code(s): Irritant

Formula: C₈H₆FN
MW: 135.14 g/mol
Boiling Pt: 90 °C (0.4mm)
Melting Pt: 30…32 °C
MDL Number: MFCD00055992
CAS Number: 387-43-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR