5-Azaindole ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 1H-pyrrolo[3,2-c]pyridine, 1H-Pyrrolo[2,3-d]pyridine

069735-1G 069735-5G 069735-25G
101941-686EA 29.23 USD
101941-686 101941-688 101941-690
5-Azaindole ≥98% for synthesis
5-Azaindole

Hazard code(s): Irritant

Formula: C₇H₆N₂
Melting Pt: 113 °C
MDL Number: MFCD00955936
CAS Number: 271-34-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR