3,4-Dihydroxy-3-cyclobutene-1,2-dione ≥99%

Supplier: Matrix Scientific

Synonyms: Squaric acid, 1,2-Dihydroxy-3,4-cyclobutenedione, Quadratic acid, 1,2-Dihydroxy-1-cyclobutene-3,4-dione, Diketocyclobutenediol

069722-25G 069722-100G
101941-616EA 49.23 USD
101941-616 101941-618
3,4-Dihydroxy-3-cyclobutene-1,2-dione ≥99%
3,4-Dihydroxy-3-cyclobutene-1,2-dione

Hazard code(s): Corrosive

Formula: C₄H₂O₄
MW: 114.06 g/mol
Melting Pt: >300 °C
Density: 1 g/cm³ (20 °C)
MDL Number: MFCD00001334
CAS Number: 2892-51-5
EINECS: 220-761-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR