4-Bromo-2-methoxypyridine ≥95%

Supplier: Matrix Scientific

069710-5G 069710-25G
101941-562EA 115.38 USD
101941-562 101941-564
4-Bromo-2-methoxypyridine ≥95%
4-Bromo-2-methoxypyridine

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₆BrNO
MDL Number: MFCD08277268
CAS Number: 100367-39-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR