4-Bromo-2-(trimethylsilyl)thiazole ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Bromo-2-trimethylsilanylthiazole

069696-1G 069696-5G 069696-25G
101941-490EA 263.08 USD
101941-490 101941-492 101941-494
4-Bromo-2-(trimethylsilyl)thiazole ≥97%
4-Bromo-2-(trimethylsilyl)thiazole

Hazard code(s): Irritant

Formula: C₆H₁₀BrNSSi
MW: 236.21 g/mol
MDL Number: MFCD01319107
CAS Number: 108306-53-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR