6-Azaindole ≥96%

Supplier: Matrix Scientific

069677-1G 069677-5G
101941-394EA 26.15 USD
101941-394 101941-396
6-Azaindole ≥96%
6-Azaindole

Hazard code(s): Irritant, Refrigerate

Formula: C₇H₆N₂
MW: 118.14 g/mol
MDL Number: MFCD01686899
CAS Number: 271-29-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR