3-Bromo-2-fluoro-4-pyridineboronic acid pinacol ester ≥96% for synthesis

Supplier: Matrix Scientific

Synonyms: 3-Bromo-2-fluoropyridine-4-boronic acid pinacol ester

069667-1G 069667-5G
101941-344EA 524.62 USD
101941-344 101941-346
3-Bromo-2-fluoro-4-pyridineboronic acid pinacol ester ≥96% for synthesis
3-Bromo-2-fluoro-4-pyridineboronic acid pinacol ester

Hazard code(s): Irritant

Formula: C₁₁H₁₄BBrFNO₂
MDL Number: MFCD08063110
CAS Number: 1150561-78-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR