2-Iodo-1-methylimidazole ≥98%
Supplier: Matrix Scientific
Synonyms:
2-Iodo-1-methyl-1H-imidazole
069651-1G
069651-5G
101941-270EA
349.23
USD
101941-270
101941-272
2-Iodo-1-methylimidazole ≥98%
2-Iodo-1-methylimidazole
Hazard code(s): Irritant
Formula:
C₄H₅IN₂ MW: 208 g/mol |
MDL Number:
MFCD00661606 CAS Number: 37067-95-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...