3-Methylphenylboronic acid pinacol ester ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Methylbenzeneboronic acid, pinacol ester, 4,4,5,5-Tetramethyl-2-(m-tolyl)-1,3,2-dioxaborolane, 3-Tolylboronic acid pinacol ester, m-Tolylboronic acid Pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)toluene

069645-5G 069645-25G 069645-100G
101941-246EA 83.08 USD
101941-246 101941-248 101941-250
3-Methylphenylboronic acid pinacol ester ≥98%
3-Methylphenylboronic acid pinacol ester

Hazard code(s): Irritant

Formula: C₁₃H₁₉BO₂
MW: 218.1 g/mol
MDL Number: MFCD05864303
CAS Number: 253342-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR