3-Amino-6-chloropyridazine ≥98%

Supplier: Matrix Scientific
Warning

069641-25G 069641-100G
101941-224EA 101.54 USD
101941-224 101941-226
3-Amino-6-chloropyridazine ≥98%
3-Amino-6-chloropyridazine

Hazard code(s): Irritant

Formula: C₄H₄ClN₃
Melting Pt: 226…228 °C
MDL Number: MFCD00051506
CAS Number: 5469-69-2
EINECS: 226-796-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR