3-Bromopyridine ≥98%

Supplier: Matrix Scientific
Warning

069610-25G 069610-100G 069610-500G
101941-068EA 16.92 USD
101941-068 101941-070 101941-072
3-Bromopyridine ≥98%
3-Bromopyridine

Hazard code(s): Flammable, Light Sensitive

Formula: C₅H₄BrN
Boiling Pt: 172…174 °C
Melting Pt: –27 °C
Density: 1.62 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00006373
CAS Number: 626-55-1
EINECS: 210-952-0
UN: 2810
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR