Methyl bromopyruvate ≥90% for synthesis

Supplier: Matrix Scientific

Synonyms: Methyl 3-Bromopyruvate, Methyl 3-Bromo-2-oxopropionate, Bromopyruvic acid methyl ester, 3-Bromopyruvic acid methyll ester, 3-Bromo-2-oxopropionic acid methyll ester, 3-Bromo-2-oxopropionic acid methyl ester

069606-5G 069606-25G 069606-100G
101941-048EA 55.38 USD
101941-048 101941-050 101941-052
Methyl bromopyruvate ≥90% for synthesis
Methyl bromopyruvate

Hazard code(s): Irritant

Formula: BrH₂CC(O)COOCH₃
Density: 1.670 g/cm³
Storage Temperature: Refrigerator
MDL Number: MFCD01862963
CAS Number: 7425-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR