2-Fluoro-4-pyridineboronic acid pinacol ester ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: 2-Fluoropyridine-4-boronic acid pinacol ester, 2-Fluoro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

069586-1G 069586-5G 069586-25G
101940-942EA 73.85 USD
101940-942 101940-944 101940-946
2-Fluoro-4-pyridineboronic acid pinacol ester ≥95% for synthesis
2-Fluoro-4-pyridineboronic acid pinacol ester

Hazard code(s): Irritant

Formula: C₁₁H₁₅BFNO₂
Melting Pt: 48…52 °C
MDL Number: MFCD06798253
CAS Number: 458532-86-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR