5-Bromo-1-methylimidazole ≥98% for synthesis

Supplier: Matrix Scientific

Synonyms: 5-Bromo-1-methyl-1H-imidazole

069530-1G 069530-5G 069530-25G
101940-640EA 263.08 USD
101940-640 101940-642 101940-644
5-Bromo-1-methylimidazole ≥98% for synthesis
5-Bromo-1-methylimidazole

Hazard code(s): Irritant

Formula: C₄H₅BrN₂
MW: 161
MDL Number: MFCD01632218
CAS Number: 1003-21-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR