2-Fluoro-3-iodopyridine ≥98%

Supplier: Matrix Scientific
Warning

069500-5G 069500-25G
101940-476EA 44.62 USD
101940-476 101940-478
2-Fluoro-3-iodopyridine ≥98%
2-Fluoro-3-iodopyridine

Hazard code(s): Irritant

Formula: C₅H₃FIN
MW: 222.99 g/mol
Melting Pt: 42…48 °C
MDL Number: MFCD03095287
CAS Number: 113975-22-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR