2,5-Diiodo-1-methylimidazole ≥98%

Supplier: Matrix Scientific

Synonyms: 2,5-Diiodo-1-methyl-1H-imidazole

069492-1G 069492-5G
101940-430EA 1221.54 USD
101940-430 101940-432
2,5-Diiodo-1-methylimidazole ≥98%
2,5-Diiodo-1-methylimidazole

Hazard code(s): Irritant

Formula: C₄H₄I₂N₂
MW: 333.9 g/mol
MDL Number: MFCD02179540
CAS Number: 86026-81-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR