5-Bromothiazole ≥98%

Supplier: Matrix Scientific

069489-1G 069489-5G 069489-25G
101940-412EA 146.15 USD
101940-412 101940-414 101940-416
5-Bromothiazole ≥98%
5-Bromothiazole

Hazard code(s): Irritant, Moisture Sensitive

Formula: C₃H₂BrNS
MW: 164.03 g/mol
Density: 1.835 g/cm³
MDL Number: MFCD07787394
CAS Number: 3034-55-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR