2-Cyanophenylboronic acid ≥95%

Supplier: Matrix Scientific
Warning

069486-5G 069486-25G
101940-396EA 113.85 USD
101940-396 101940-398
2-Cyanophenylboronic acid ≥95%
2-Cyanophenylboronic acid

Hazard code(s): Irritant

Formula: C₇H₆BNO₂
MDL Number: MFCD01632208
CAS Number: 138642-62-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR