3-Chloro-4-pyridineboronic acid ≥96%

Supplier: Matrix Scientific

069484-1G 069484-5G
101940-386EA 3670.77 USD
101940-386 101940-388
3-Chloro-4-pyridineboronic acid ≥96%
3-Chloro-4-pyridineboronic acid

Hazard code(s): Irritant

Formula: C₅H₅BClNO₂
MW: 157.36 g/mol
MDL Number: MFCD03425942
CAS Number: 458532-98-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR