5-Bromo-2-methoxypyrimidine ≥98%
Supplier: Matrix Scientific
069463-1G
069463-5G
069463-25G
101940-266EA
58.46
USD
101940-266
101940-268
101940-270
5-Bromo-2-methoxypyrimidine ≥98%
5-Bromo-2-methoxypyrimidine
Hazard code(s): Irritant
Formula:
C₅H₅BrN₂O MW: 189.01 g/mol |
MDL Number:
MFCD04973351 CAS Number: 14001-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...