4-Iodo-1-methylimidazole ≥97%

Supplier: Matrix Scientific

Synonyms: 4-Iodo-1-methyl-1H-imidazole

069445-1G 069445-5G
101940-160EA 113.85 USD
101940-160 101940-162
4-Iodo-1-methylimidazole ≥97%
4-Iodo-1-methylimidazole

Hazard code(s): Irritant

Formula: C₄H₅IN₂
MW: 208 g/mol
MDL Number: MFCD01632214
CAS Number: 71759-87-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR