2-Bromo-5-thiazolecarboxylic acid ≥97%

Supplier: Matrix Scientific

069440-5G 069440-25G
101940-136EA 49.23 USD
101940-136 102551-750
2-Bromo-5-thiazolecarboxylic acid ≥97%

Hazard code(s): Irritant

Formula: C₄H₂BrNO₂S
Melting Pt: 180…184 °C
MDL Number: MFCD04115730
CAS Number: 54045-76-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR