2-Bromo-4-thiazolecarboxylic acid ≥97%
Supplier: Matrix Scientific
069439-1G
069439-5G
101940-128EA
78.46
USD
101940-128
101940-130
2-Bromo-4-thiazolecarboxylic acid ≥97%
2-Bromo-4-thiazolecarboxylic acid
Hazard code(s): Irritant
Formula:
C₄H₂BrNO₂S |
MDL Number:
MFCD04115729 CAS Number: 5198-88-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...