2-Bromoquinoline ≥98%

Supplier: Matrix Scientific
Warning

069148-1G 069148-5G 069148-25G
101939-086EA 233.85 USD
101939-086 101939-088 101939-090
2-Bromoquinoline ≥98%
2-Bromoquinoline

Hazard code(s): Irritant

Formula: C₉H₆BrN
MW: 208.06 g/mol
MDL Number: MFCD00234480
CAS Number: 2005-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR