5-Bromoquinoline ≥98%
Supplier: Matrix Scientific
|
Warning
|
069144-5G
069144-25G
101939-074EA
610.77
USD
101939-074
101939-076
5-Bromoquinoline ≥98%
5-Bromoquinoline
Hazard code(s): Irritant
Formula:
C₉H₆BrN MW: 208.06 g/mol |
MDL Number:
MFCD00234481 CAS Number: 4964-71-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...