2-Chloro-6-methyl-3-nitropyridine ≥98%

Supplier: Matrix Scientific

069086-1G 069086-5G 069086-25G
101938-870EA 26.15 USD
101938-870 101938-872 101938-874
2-Chloro-6-methyl-3-nitropyridine ≥98%
2-Chloro-6-methyl-3-nitropyridine

Hazard code(s): Irritant, Store in Freezer

Formula: C₆H₅ClN₂O₂
MDL Number: MFCD03085820
CAS Number: 56057-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR