4-(Dimethylamino)benzoyl chloride ≥95%

Supplier: Matrix Scientific

068970-1G 068970-5G
101938-474EA 87.69 USD
101938-474 101938-476
4-(Dimethylamino)benzoyl chloride ≥95%
4-(Dimethylamino)benzoyl chloride

Hazard code(s): Corrosive, Moisture Sensitive

Formula: C₉H₁₀ClNO
MW: 183.64 g/mol
Boiling Pt: 180 °C (15 -rr)
Melting Pt: 147 - 148 °C
MDL Number: MFCD00236039
CAS Number: 4755-50-4
UN: 3261
ADR: 8,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR