4(3H)-Pyrimidinone ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Pyrimidinol, 4(3H)-Pyrimidone, 3H-Pyrimidin-4-one, 4-Hydroxypyrimidine, Pyrimidin-4(3H)-one

068598-5G 068598-25G
101936-852EA 100 USD
101936-852 101936-854
4(3H)-Pyrimidinone ≥95%
4(3H)-Pyrimidinone

Hazard code(s): Irritant

Formula: C₄H₄N₂O
MW: 96.09 g/mol
MDL Number: MFCD00006664
CAS Number: 4562-27-0
EINECS: 224-932-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR