1-Methyl-1H-pyrazole-5-boronic acid ≥95% for synthesis

Supplier: Matrix Scientific

Synonyms: (1-Methyl-1H-pyrazol-5-yl)boronic acid, (1-Methyl-1H-pyrazol-5-yl)-boronic acid

068538-1G 068538-5G
101936-584EA 30.77 USD
101936-584 101936-586
1-Methyl-1H-pyrazole-5-boronic acid ≥95% for synthesis
1-Methyl-1H-pyrazole-5-boronic acid

Hazard code(s): Irritant

Formula: C₄H₇BN₂O₂
MDL Number: MFCD11053031
CAS Number: 720702-41-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR