Quinuclidin-3-ona ≥95%

Supplier: Matrix Scientific

Synonyms: 1-Azabicyclo[2.2.2]octan-3-one

068491-1G 068491-5G
101936-378EA 136.92 USD
101936-378 101936-380
Quinuclidin-3-ona ≥95%
Quinuclidin-3-ona

Hazard code(s): Irritant

Formula: C₇H₁₁NO
Melting Pt: >300 °C
MDL Number: MFCD06801385
CAS Number: 3731-38-2
EINECS: 223-087-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR