2-(Boc-amino)pyridine ≥95%

Supplier: Matrix Scientific

Synonyms: tert-Butyl N-(2-pyridyl)carbamate, tert-Butyl pyridin-2-ylcarbamate

068387-1G 068387-5G
101935-908EA 233.85 USD
101935-908 101935-910
2-(Boc-amino)pyridine ≥95%
2-(Boc-amino)pyridine

Hazard code(s): Irritant

Formula: C₁₀H₁₄N₂O₂
MW: 194.23 g/mol
MDL Number: MFCD03411622
CAS Number: 38427-94-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR