4-Bromo-2,3-difluoroanisole ≥98%
Supplier: Matrix Scientific
067253-1G
067253-5G
067253-25G
101933-632EA
2307.69
USD
101933-632
101933-634
101933-636
4-Bromo-2,3-difluoroanisole ≥98%
4-Bromo-2,3-difluoroanisole
Hazard code(s): Irritant
Formula:
C₇H₅BrF₂O Boiling Pt: 203 °C Density: 1.615 g/cm³ |
MDL Number:
MFCD07368017 CAS Number: 406482-22-2 |
Specification Test Results
Boiling point | 203°C |
Density | 1.615 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...