5-Bromo-3-nitro-2-picoline ≥98% for synthesis
Supplier: Matrix Scientific
Synonyms:
5-Bromo-2-methyl-3-nitropyridine
067245-5G
067245-25G
101933-624EA
29.23
USD
101933-624
101933-626
5-Bromo-3-nitro-2-picoline ≥98% for synthesis
5-Bromo-3-nitro-2-picoline
Hazard code(s): Irritant
Formula:
C₆H₅BrN₂O₂ |
MDL Number:
MFCD09031419 CAS Number: 911434-05-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...