3-Bromo-4-formylpyridine ≥96%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-4-pyridinecarboxaldehyde, 3-Bromoisonicotinaldehyde, 3-Bromopyridine-4-carboxaldehyde

067232-1G 067232-5G
101933-596EA 29.23 USD
101933-596 101933-598
3-Bromo-4-formylpyridine ≥96%
3-Bromo-4-formylpyridine

Hazard code(s): Irritant, Refrigerate

Formula: C₆H₄BrNO
MW: 186.01 g/mol
Melting Pt: 81…82 °C
MDL Number: MFCD05864506
CAS Number: 70201-43-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR