Methyl-2-amino-6-chloronicotinate
Supplier: Matrix Scientific
Synonyms:
Methyl 2-amino-6-chloronicotinate
067189-500MG
067189-1G
101933-446EA
361.54
USD
101933-446
101933-448
Methyl-2-amino-6-chloronicotinate
Methyl-2-amino-6-chloronicotinate
Hazard code(s): Irritant
Formula:
C₇H₇ClN₂O₂ MW: 186.6 g/mol Melting Pt: 141…143 °C |
MDL Number:
MFCD17015967 CAS Number: 849805-25-0 |
Specification Test Results
Melting point | 141-143°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...