2-Fluoro-5-methoxybenzonitrile ≥97%
Supplier: Matrix Scientific
Synonyms:
1-Cyan-2-fluor-5-methoxybenzene
067115-1G
067115-5G
101933-192EA
29.23
USD
101933-192
101933-194
2-Fluoro-5-methoxybenzonitrile ≥97%
2-Fluoro-5-methoxybenzonitrile
Hazard code(s): Irritant
Formula:
C₈H₆FNO MW: 151.14 g/mol |
MDL Number:
MFCD00142921 CAS Number: 127667-01-0 UN: 3439 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...