4-Bromo-1-methylimidazole ≥97%
Supplier: Matrix Scientific
Synonyms:
4-Bromo-1-methyl-1H-imidazole
067109-1G
067109-5G
101933-180EA
100
USD
101933-180
102833-424
4-Bromo-1-methylimidazole ≥97%
4-Bromo-1-methylimidazole
Hazard code(s): Irritant
Formula:
C₄H₅BrN₂ MW: 161 g/mol |
MDL Number:
MFCD01320501 CAS Number: 25676-75-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...