1-Bromo-2-chloro-4-fluorobenzene ≥98%

Supplier: Matrix Scientific
Warning

Synonyms: 4-Bromo-3-chloro-1-fluorobenzene, 2-Chloro-4-fluorobromobenzene, 4-Bromo-3-chlorofluorobenzene

067064-5G 067064-25G 067064-100G
101933-108EA 20 USD
101933-108 101933-110 102551-692
1-Bromo-2-chloro-4-fluorobenzene ≥98%
1-Bromo-2-chloro-4-fluorobenzene

Hazard code(s): Irritant

Formula: C₆H₃BrClF
MW: 209.44 g/mol
MDL Number: MFCD00142587
CAS Number: 110407-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR