2-Bromo-4-chloro-1-fluorobenzene ≥98%

Supplier: Matrix Scientific

Synonyms: 3-Bromo-4-fluorochlorobenzene, 2-Bromo-4-chlorofluorobenzene, 1-Bromo-3-chloro-6-fluorobenzene, 3-Bromo-1-chloro-4-fluorobenzene

067063-5G 067063-25G 067063-100G
101933-102EA 15.38 USD
101933-102 101933-104 101933-106
2-Bromo-4-chloro-1-fluorobenzene ≥98%
2-Bromo-4-chloro-1-fluorobenzene

Hazard code(s): Irritant

Formula: C₆H₃BrClF
MW: 209.44 g/mol
MDL Number: MFCD00672934
CAS Number: 1996-30-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR