4-Bromo-3-chlorobenzonitrile ≥98%
Supplier: Matrix Scientific
066951-5G
066951-25G
101932-850EA
44.62
USD
101932-850
101932-852
4-Bromo-3-chlorobenzonitrile ≥98%
4-Bromo-3-chlorobenzonitrile
Hazard code(s): Irritant
Formula:
C₇H₃BrClN MW: 216.46 g/mol Boiling Pt: 261 °C Melting Pt: 80…81 °C Density: 1.74 g/cm³ |
MDL Number:
MFCD09834777 CAS Number: 57418-97-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...