3-Bromo-4-chloroanisole ≥98%

Supplier: Matrix Scientific

Synonyms: 2-Bromo-1-chloro-4-methoxybenzene

066949-5G 066949-25G
101932-842EA 21.54 USD
101932-842 101932-844
3-Bromo-4-chloroanisole ≥98%
3-Bromo-4-chloroanisole

Hazard code(s): Irritant

Formula: C₇H₆BrClO
MW: 221.48 g/mol
MDL Number: MFCD00070739
CAS Number: 2732-80-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR