2-Bromo-5-chloroanisole ≥98%
Supplier: Matrix Scientific
Synonyms:
1-Bromo-4-chloro-2-methoxybenzene
066948-5G
066948-25G
101932-838EA
30.77
USD
101932-838
101932-840
2-Bromo-5-chloroanisole ≥98%
2-Bromo-5-chloroanisole
Hazard code(s): Irritant
Formula:
C₇H₆BrClO MW: 221.48 g/mol |
MDL Number:
MFCD03790889 CAS Number: 174913-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...