3-Bromo-5-chloro-2-methoxypyridine ≥98%
Supplier: Matrix Scientific
066866-1G
066866-5G
101932-552EA
23.08
USD
101932-552
101932-554
3-Bromo-5-chloro-2-methoxypyridine ≥98%
3-Bromo-5-chloro-2-methoxypyridine
Hazard code(s): Irritant
Formula:
C₆H₅BrClNO Melting Pt: 48…50 °C |
MDL Number:
MFCD08059322 CAS Number: 102830-75-1 |
Specification Test Results
Melting point | 48-50°C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...