3-Bromo-2-nitroaniline ≥98%

Supplier: Matrix Scientific

066860-1G 066860-5G 066860-25G
101932-540EA 15.38 USD
101932-540 101932-542 102663-826
3-Bromo-2-nitroaniline ≥98%
3-Bromo-2-nitroaniline

Hazard code(s): Irritant

Formula: C₆H₅BrN₂O₂
MW: 217.02
MDL Number: MFCD08063248
CAS Number: 7138-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR