5-Aminopicolinic acid ≥97%

Supplier: Matrix Scientific

Synonyms: 5-Aminopyridine-2-carboxylic acid, 5-Amino-2-pyridinecarboxylic acid

066857-1G 066857-5G
101932-536EA 26.15 USD
101932-536 101932-538
5-Aminopicolinic acid ≥97%
5-Aminopicolinic acid

Hazard code(s): Irritant

Formula: C₆H₆N₂O₂
MW: 138.13 g/mol
Melting Pt: 217…219 °C
MDL Number: MFCD02684600
CAS Number: 24242-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR