Benzo[b]thiophene ≥97%

Supplier: Matrix Scientific

Synonyms: Thianaphthene, 1-Benzothiophene

066631-25G 066631-100G
101932-024EA 92.31 USD
101932-024 101932-026
Benzo[b]thiophene ≥97%
Benzo[b]thiophene

Hazard code(s): Irritant

Formula: C₈H₆S
MW: 134.2 g/mol
Boiling Pt: 221…222 °C
Melting Pt: 28…32 °C
Density: 1.149 g/cm³
MDL Number: MFCD00005864
CAS Number: 95-15-8
EINECS: 202-395-7

Order Now

Specification Test Results

Boiling point 221-222°C
Melting point 28-32°C
Density 1.149

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR