N-Benzylmaleimide for synthesis

Supplier: Matrix Scientific

Synonyms: 1-Benzyl-1H-pyrrole-2,5-dione, 1-Benzylpyrrole-2,5-dione

066498-5G 066498-25G
101931-774EA 18.46 USD
101931-774 101931-776
N-Benzylmaleimide for synthesis
N-Benzylmaleimide
Formula: C₁₁H₉NO₂
MW: 187.2 g/mol
Boiling Pt: 339
Melting Pt: 70…72 °C
Density: 1.296 g/cm³ (20 °C)
MDL Number: MFCD00014540
CAS Number: 1631-26-1
EINECS: 216-631-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR